siRNA Details
| Gene Symbol: Lamc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100640.1 | NP_001094110.1 | 20, 21 | 3172 | ||
| XM_008769602.1 | XP_008767824.1 | 19, 20 | 3134 | |||
| XM_008769603.1 | XP_008767825.1 | 20, 21 | 3010 | |||

Lamc2
Genomic Map