siRNA Details
| Gene Symbol: Mtnr1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100641.1 | NP_001094111.1 | 2 | 922 | ||
| GenBank mRNA | AB377275.1 | - | 2 | 922 | ||
| AF141863.1 | - | 1 | 393 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mtnr1b| Gene Symbol: Mtnr1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100641.1 | NP_001094111.1 | 2 | 922 | ||
| GenBank mRNA | AB377275.1 | - | 2 | 922 | ||
| AF141863.1 | - | 1 | 393 | |||
Set Membership: |
|