siRNA Details
| Gene Symbol: Hist1h1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133285.1 | NP_579819.1 | 1 | 431 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hist1h1d| Gene Symbol: Hist1h1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133285.1 | NP_579819.1 | 1 | 431 | ||