siRNA Details
| Gene Symbol: Asip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052979.1 | NP_443211.1 | 2, 3 | 245 | ||
| GenBank mRNA | AB045587.1 | - | 2, 3 | 245 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Asip| Gene Symbol: Asip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052979.1 | NP_443211.1 | 2, 3 | 245 | ||
| GenBank mRNA | AB045587.1 | - | 2, 3 | 245 | ||