siRNA Details
| Gene Symbol: Ak1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024349.3 | NP_077325.2 | 558 | |||
| GenBank mRNA | AB022701.1 | - | 520 | |||
| BC089797.1 | - | 558 | 7130672 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ak1| Gene Symbol: Ak1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024349.3 | NP_077325.2 | 558 | |||
| GenBank mRNA | AB022701.1 | - | 520 | |||
| BC089797.1 | - | 558 | 7130672 | |||