siRNA Details
| Gene Symbol: Ascl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031503.2 | NP_113691.1 | 2 | 713 | ||
| GenBank mRNA | X53724.1 | - | 2 | 657 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ascl2| Gene Symbol: Ascl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031503.2 | NP_113691.1 | 2 | 713 | ||
| GenBank mRNA | X53724.1 | - | 2 | 657 | ||