siRNA Details
Gene Symbol: Avp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016992.2 | NP_058688.2 | 3 | 490 | ||
GenBank mRNA | M25646.1 | - | 3 | 490 | ||
M64785.1 | - | 838 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Avp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016992.2 | NP_058688.2 | 3 | 490 | ||
GenBank mRNA | M25646.1 | - | 3 | 490 | ||
M64785.1 | - | 838 |