siRNA Details
| Gene Symbol: Tspo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012515.2 | NP_036647.1 | 4 | 541 | ||
| GenBank mRNA | BQ194961.1 | - | 1 | 313 | ||
| FQ228784.1 | - | 4 | 543 | |||
| J05122.1 | - | 3 | 484 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tspo| Gene Symbol: Tspo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012515.2 | NP_036647.1 | 4 | 541 | ||
| GenBank mRNA | BQ194961.1 | - | 1 | 313 | ||
| FQ228784.1 | - | 4 | 543 | |||
| J05122.1 | - | 3 | 484 | |||