siRNA Details
| Gene Symbol: Calm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031969.2 | NP_114175.1 | 3 | 167 | ||
| GenBank mRNA | AF178845.1 | - | 102 | |||
| FQ211760.1 | - | 3 | 234 | |||
| FQ212721.1 | - | 572 | ||||
| FQ213227.1 | - | 3 | 280 | |||
| FQ225380.1 | - | 3 | 193 | |||
| FQ226971.1 | - | 3 | 239 | |||
| FQ227186.1 | - | 3 | 239 | |||
| X13933.1 | - | 3 | 167 | |||

Calm1
Genomic Map