siRNA Details
| Gene Symbol: Calm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012518.3 | NP_036650.1 | 3 | 227 | ||
| GenBank mRNA | BC063187.1 | - | 322 | 6890866 | ||
| FQ212716.1 | - | 293 | ||||
| M16659.1 | - | 3 | 227 | |||
| X13817.1 | - | 4 | 319 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Calm3| Gene Symbol: Calm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012518.3 | NP_036650.1 | 3 | 227 | ||
| GenBank mRNA | BC063187.1 | - | 322 | 6890866 | ||
| FQ212716.1 | - | 293 | ||||
| M16659.1 | - | 3 | 227 | |||
| X13817.1 | - | 4 | 319 | |||