siRNA Details
| Gene Symbol: Cgm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012525.1 | NP_036657.1 | 4 | 1177 | ||
| GenBank mRNA | BC097366.1 | - | 4 | 1141 | 7443149 | |
| BI285964.1 | - | 2 | 573 | |||
| CB766157.1 | - | 2 | 357 | |||
| CB786391.1 | - | 1 | 128 | |||

Cgm4
Genomic Map