siRNA Details
| Gene Symbol: Chga | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021655.2 | NP_067687.2 | 2 | 213 | ||
| GenBank mRNA | AF145445.1 | - | 2 | 75 | ||
| BC087703.1 | - | 2 | 193 | 7313842 | ||
| CB743841.1 | - | 2 | 269 | |||
| X06832.1 | - | 2 | 198 | |||

Chga
Genomic Map