siRNA Details
Gene Symbol: Chga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021655.2 | NP_067687.2 | 3 | 273 | ||
GenBank mRNA | AF145445.1 | - | 3 | 135 | ||
BC087703.1 | - | 3 | 253 | 7313842 | ||
CB743841.1 | - | 3 | 329 | |||
X06832.1 | - | 3 | 258 |
