siRNA Details
| Gene Symbol: Chga | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021655.2 | NP_067687.2 | 4 | 377 | ||
| GenBank mRNA | AF145445.1 | - | 4 | 239 | ||
| BC087703.1 | - | 4 | 357 | 7313842 | ||
| X06832.1 | - | 4 | 362 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chga| Gene Symbol: Chga | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021655.2 | NP_067687.2 | 4 | 377 | ||
| GenBank mRNA | AF145445.1 | - | 4 | 239 | ||
| BC087703.1 | - | 4 | 357 | 7313842 | ||
| X06832.1 | - | 4 | 362 | |||