siRNA Details
| Gene Symbol: Cp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270961.1 | NP_001257890.1 | 16 | 2712 | ||
| NM_012532.2 | NP_036664.2 | 16 | 2712 | |||
| XM_008760859.1 | XP_008759081.1 | 16 | 2900 | |||
| GenBank mRNA | AF202115.1 | - | 16 | 2679 | ||
| FQ220241.1 | - | 3 | 229 | |||
| L33869.1 | - | 16 | 2694 | |||

Cp
Genomic Map