siRNA Details
| Gene Symbol: Egfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031507.1 | NP_113695.1 | 23 | 2915 | ||
| XM_008770416.1 | XP_008768638.1 | 23 | 2994 | |||
| XM_008770417.1 | XP_008768639.1 | 22 | 2952 | |||
| GenBank mRNA | AB079120.1 | - | 3 | 185 | ||
| HM801041.1 | - | 23 | 2880 | |||
| HM801042.1 | - | 23 | 2880 | |||
| M37394.2 | - | 23 | 2915 | |||

Egfr
Genomic Map