siRNA Details
| Gene Symbol: Egfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031507.1 | NP_113695.1 | 25 | 3184 | ||
| XM_008770416.1 | XP_008768638.1 | 25 | 3263 | |||
| XM_008770417.1 | XP_008768639.1 | 24 | 3221 | |||
| GenBank mRNA | AB079120.1 | - | 5 | 454 | ||
| HM801041.1 | - | 25 | 3149 | |||
| HM801042.1 | - | 25 | 3149 | |||
| M37394.2 | - | 25 | 3184 | |||

Egfr
Genomic Map