siRNA Details
| Gene Symbol: Egfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031507.1 | NP_113695.1 | 25, 26 | 3275 | ||
| XM_008770416.1 | XP_008768638.1 | 25, 26 | 3354 | |||
| XM_008770417.1 | XP_008768639.1 | 24, 25 | 3312 | |||
| GenBank mRNA | AB079120.1 | - | 5, 6 | 545 | ||
| HM801041.1 | - | 25, 26 | 3240 | |||
| HM801042.1 | - | 25, 26 | 3240 | |||
| M37394.2 | - | 25, 26 | 3275 | |||
| U52529.1 | - | 1, 2 | 33 | |||

Egfr
Genomic Map