siRNA Details
| Gene Symbol: Fgg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012559.2 | NP_036691.2 | 6 | 640 | ||
| XM_006232525.1 | XP_006232587.1 | 6 | 794 | |||
| GenBank mRNA | BC078893.1 | - | 6 | 640 | 7122366 | |
| FQ209468.1 | - | 2 | 113 | |||
| FQ218666.1 | - | 4 | 442 | |||
| FQ219408.1 | - | 6 | 660 | |||
| FQ219511.1 | - | 6 | 662 | |||
| FQ219541.1 | - | 6 | 661 | |||
| FQ219604.1 | - | 6 | 661 | |||
| FQ229132.1 | - | 6 | 667 | |||

Fgg
Genomic Map