siRNA Details
| Gene Symbol: Gfap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017009.2 | NP_058705.2 | 8, 9 | 1245 | ||
| GenBank mRNA | BC088851.1 | - | 8, 9 | 1245 | 7315338 | |
| L27219.1 | - | 8, 9 | 1249 | |||
| U03700.1 | - | 8, 9 | 1269 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gfap| Gene Symbol: Gfap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017009.2 | NP_058705.2 | 8, 9 | 1245 | ||
| GenBank mRNA | BC088851.1 | - | 8, 9 | 1245 | 7315338 | |
| L27219.1 | - | 8, 9 | 1249 | |||
| U03700.1 | - | 8, 9 | 1269 | |||