siRNA Details
| Gene Symbol: Gls | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109968.1 | NP_001103438.1 | 12, 13 | 1494 | ||
| NM_012569.2 | NP_036701.2 | 12, 13 | 1494 | |||
| XM_008767075.1 | XP_008765297.1 | 12, 13 | 1665 | |||
| GenBank mRNA | AY083459.1 | - | 7, 8 | 549 | ||
| CB776615.1 | - | 4, 5 | 323 | |||
| M22586.1 | - | 5, 6 | 399 | |||
| M65150.1 | - | 12, 13 | 1501 | |||

Gls
Genomic Map