siRNA Details
| Gene Symbol: Got1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012571.2 | NP_036703.2 | 3, 4 | 505 | ||
| GenBank mRNA | BC061877.1 | - | 3, 4 | 478 | 5599273 | |
| CB578328.1 | - | 1, 2 | 69 | |||
| D00252.1 | - | 3, 4 | 462 | |||
| FM055803.1 | - | 3, 4 | 427 | |||
| J04171.1 | - | 3, 4 | 446 | |||

Got1
Genomic Map