siRNA Details
| Gene Symbol: Gpx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030826.3 | NP_110453.3 | 1, 2 | 313 | ||
| GenBank mRNA | BC058438.1 | - | 1, 2 | 285 | 6918849 | |
| BI281978.1 | - | 1, 2 | 582 | |||
| CB579218.1 | - | 1, 2 | 313 | |||
| FQ211052.1 | - | 1, 2 | 310 | |||
| FQ227523.1 | - | 1, 2 | 342 | |||
| M21210.1 | - | 1, 2 | 319 | |||

Gpx1
Genomic Map