siRNA Details
| Gene Symbol: Gpx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030826.3 | NP_110453.3 | 2 | 374 | ||
| GenBank mRNA | BC058438.1 | - | 2 | 346 | 6918849 | |
| BI281978.1 | - | 1 | 521 | |||
| CB579218.1 | - | 2 | 374 | |||
| FQ211052.1 | - | 2 | 371 | |||
| FQ227523.1 | - | 2 | 403 | |||
| M21210.1 | - | 2 | 380 | |||

Gpx1
Genomic Map