siRNA Details
| Gene Symbol: Hgf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017017.2 | NP_058713.1 | 7 | 989 | ||
| XM_006235979.2 | XP_006236041.1 | 7 | 1069 | |||
| GenBank mRNA | D90102.1 | - | 7 | 989 | ||
| X54400.1 | - | 7 | 888 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hgf| Gene Symbol: Hgf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017017.2 | NP_058713.1 | 7 | 989 | ||
| XM_006235979.2 | XP_006236041.1 | 7 | 1069 | |||
| GenBank mRNA | D90102.1 | - | 7 | 989 | ||
| X54400.1 | - | 7 | 888 | |||