siRNA Details
| Gene Symbol: Ibsp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012587.2 | NP_036719.2 | 5 | 295 | ||
| GenBank mRNA | AB001383.1 | - | 4 | 222 | ||
| BC127506.1 | - | 5 | 291 | 8369751 | ||
| DQ213013.1 | - | 4 | 222 | |||
| DY314698.1 | - | 5 | 295 | |||
| J04215.1 | - | 5 | 301 | |||

Ibsp
Genomic Map