siRNA Details
| Gene Symbol: Ibsp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012587.2 | NP_036719.2 | 6, 7 | 482 | ||
| GenBank mRNA | AB001383.1 | - | 5, 6 | 409 | ||
| BC127506.1 | - | 6, 7 | 478 | 8369751 | ||
| DQ213013.1 | - | 5, 6 | 409 | |||
| DY313346.1 | - | 1, 2 | 158 | |||
| DY314698.1 | - | 6, 7 | 482 | |||
| J04215.1 | - | 6, 7 | 488 | |||

Ibsp
Genomic Map