siRNA Details
| Gene Symbol: Idh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031510.1 | NP_113698.1 | 7 | 1111 | ||
| XM_006245049.2 | XP_006245111.1 | 8 | 1169 | |||
| XM_008767076.1 | XP_008765298.1 | 8 | 1201 | |||
| GenBank mRNA | DQ403076.1 | - | 7 | 1001 | ||
| FQ214242.1 | - | 8 | 1192 | |||
| L35317.1 | - | 7 | 1111 | |||

Idh1
Genomic Map