siRNA Details
Gene Symbol: Igf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082477.2 | NP_001075946.2 | 5 | 475 | ||
NM_001082478.1 | NP_001075947.1 | 5 | 772 | |||
NM_001082479.1 | NP_001075948.1 | 4 | 720 | |||
NM_178866.4 | NP_849197.3 | 4 | 423 | |||
XM_006241191.2 | XP_006241253.1 | 6 | 616 | |||
XM_006241192.1 | XP_006241254.1 | 6 | 530 | |||
XM_006241193.2 | XP_006241255.1 | 5 | 563 | |||
XM_006241194.2 | XP_006241256.1 | 5 | 654 | |||
GenBank mRNA | BC086374.1 | - | 4 | 423 | 7300903 | |
D00698.1 | - | 4 | 404 | |||
FQ209824.1 | - | 5 | 552 | |||
FQ210560.1 | - | 5 | 488 | |||
FQ210688.1 | - | 4 | 436 | |||
FQ220385.1 | - | 5 | 796 | |||
FQ225388.1 | - | 4 | 436 | |||
FQ225437.1 | - | 5 | 436 | |||
FQ231951.1 | - | 4 | 436 | |||
FQ234943.1 | - | 4 | 436 | |||
M15480.1 | - | 5 | 519 | |||
M15481.1 | - | 4 | 1155 | |||
M17335.1 | - | 4 | 620 | |||
X06043.1 | - | 3 | 305 | |||
X06107.1 | - | 5 | 750 | |||
X06108.1 | - | 6 | 564 |
