siRNA Details
Gene Symbol: Igf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190162.1 | NP_001177091.1 | 2 | 115 | ||
NM_001190163.1 | NP_001177092.1 | 2 | 103 | |||
NM_031511.2 | NP_113699.2 | 2 | 1090 | |||
XM_006230664.2 | XP_006230726.1 | 2 | 892 | |||
XM_006230665.2 | XP_006230727.1 | 2 | 95 | |||
XM_008760074.1 | XP_008758296.1 | 2 | 899 | |||
XM_008760075.1 | XP_008758297.1 | 3 | 428 | |||
GenBank mRNA | JQ408382.1 | - | 1028 | |||
X00911.1 | - | 2 | 1090 | |||
X13101.1 | - | 2 | 127 | |||
X14833.1 | - | 2 | 403 | |||
X14834.1 | - | 2 | 1158 |
