siRNA Details
| Gene Symbol: Igfbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012588.2 | NP_036720.2 | 2 | 543 | ||
| GenBank mRNA | M31837.1 | - | 2 | 496 | ||
| M33300.1 | - | 2 | 473 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Igfbp3| Gene Symbol: Igfbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012588.2 | NP_036720.2 | 2 | 543 | ||
| GenBank mRNA | M31837.1 | - | 2 | 496 | ||
| M33300.1 | - | 2 | 473 | |||