siRNA Details
| Gene Symbol: Igfbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012588.2 | NP_036720.2 | 4 | 921 | ||
| GenBank mRNA | BF281457.1 | - | 3 | 384 | ||
| M31837.1 | - | 4 | 874 | |||
| M33300.1 | - | 4 | 851 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Igfbp3| Gene Symbol: Igfbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012588.2 | NP_036720.2 | 4 | 921 | ||
| GenBank mRNA | BF281457.1 | - | 3 | 384 | ||
| M31837.1 | - | 4 | 874 | |||
| M33300.1 | - | 4 | 851 | |||