siRNA Details
| Gene Symbol: Kcnj1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017023.1 | NP_058719.1 | 2 | 232 | ||
| GenBank mRNA | AF081365.1 | - | 2 | 232 | ||
| AF081366.1 | - | 2 | 181 | |||
| AF081367.1 | - | 3 | 199 | |||
| AF081368.1 | - | 3 | 354 | |||
| L29403.1 | - | 2 | 181 | |||
| X72341.1 | - | 2 | 232 | |||

Kcnj1
Genomic Map