siRNA Details
| Gene Symbol: Klk1c7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012593.1 | NP_036725.1 | 2 | 74 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klk1c7| Gene Symbol: Klk1c7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012593.1 | NP_036725.1 | 2 | 74 | ||