siRNA Details
Gene Symbol: Lalba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012594.1 | NP_036726.1 | 2 | 176 | ||
GenBank mRNA | V01251.1 | - | 1 | 52 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lalba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012594.1 | NP_036726.1 | 2 | 176 | ||
GenBank mRNA | V01251.1 | - | 1 | 52 |