siRNA Details
| Gene Symbol: Lalba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012594.1 | NP_036726.1 | 2 | 275 | ||
| XM_008765668.1 | XP_008763890.1 | 3 | 2864 | |||
| GenBank mRNA | V01251.1 | - | 1 | 151 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lalba| Gene Symbol: Lalba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012594.1 | NP_036726.1 | 2 | 275 | ||
| XM_008765668.1 | XP_008763890.1 | 3 | 2864 | |||
| GenBank mRNA | V01251.1 | - | 1 | 151 | ||