siRNA Details
Gene Symbol: Mog | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022668.2 | NP_073159.2 | 6 | 721 | ||
XM_006255867.2 | XP_006255929.1 | 6 | 1194 | |||
XM_008772687.1 | XP_008770909.1 | 6 | 1198 | |||
GenBank mRNA | BC087717.1 | - | 6 | 721 | 7317044 | |
FQ214199.1 | - | 6 | 701 | |||
L21995.1 | - | 5 | 552 | |||
M99485.1 | - | 6 | 721 |
