siRNA Details
| Gene Symbol: Myh11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170600.1 | NP_001164071.1 | 29 | 4087 | ||
| XM_006245842.2 | XP_006245904.1 | 31 | 4215 | |||
| XM_006245844.2 | XP_006245906.1 | 31 | 4210 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Myh11| Gene Symbol: Myh11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170600.1 | NP_001164071.1 | 29 | 4087 | ||
| XM_006245842.2 | XP_006245904.1 | 31 | 4215 | |||
| XM_006245844.2 | XP_006245906.1 | 31 | 4210 | |||