siRNA Details
| Gene Symbol: Cst11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139085.1 | NP_620785.1 | 1 | 46 | ||
| GenBank mRNA | AF501290.1 | - | 1 | 46 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cst11| Gene Symbol: Cst11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139085.1 | NP_620785.1 | 1 | 46 | ||
| GenBank mRNA | AF501290.1 | - | 1 | 46 | ||