siRNA Details
| Gene Symbol: Sycn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139086.1 | NP_620786.1 | 1 | 306 | ||
| GenBank mRNA | AF008197.1 | - | 1 | 306 | ||
| AF012887.1 | - | 286 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sycn| Gene Symbol: Sycn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139086.1 | NP_620786.1 | 1 | 306 | ||
| GenBank mRNA | AF008197.1 | - | 1 | 306 | ||
| AF012887.1 | - | 286 | ||||