siRNA Details
Gene Symbol: Scaf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037347.2 | NP_001032424.2 | 13 | 2126 | ||
XM_006248024.2 | XP_006248086.1 | 13 | 2240 | |||
XM_006248025.2 | XP_006248087.1 | 13 | 2240 | |||
XM_008768558.1 | XP_008766780.1 | 13 | 2228 | |||
GenBank mRNA | AY387086.1 | - | 13 | 1718 | ||
U49058.1 | - | 12 | 2001 |
