siRNA Details
Gene Symbol: Atp5e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139099.1 | NP_620799.1 | 2 | 185 | ||
GenBank mRNA | AF010323.1 | - | 2 | 185 | ||
BC058133.1 | - | 2 | 175 | 6887013 | ||
FQ211630.1 | - | 2 | 207 | |||
FQ211898.1 | - | 2 | 219 | |||
FQ214112.1 | - | 2 | 207 | |||
FQ214935.1 | - | 2 | 210 | |||
FQ215084.1 | - | 2 | 137 | |||
FQ216950.1 | - | 2 | 207 | |||
FQ217027.1 | - | 2 | 171 | |||
FQ217160.1 | - | 2 | 161 | |||
FQ217179.1 | - | 2 | 210 | |||
FQ217559.1 | - | 2 | 160 | |||
FQ217635.1 | - | 2 | 166 | |||
FQ218023.1 | - | 2 | 166 | |||
FQ218046.1 | - | 2 | 166 | |||
FQ222358.1 | - | 2 | 194 | |||
FQ223590.1 | - | 2 | 219 | |||
FQ223710.1 | - | 2 | 166 | |||
FQ223763.1 | - | 2 | 207 | |||
FQ223780.1 | - | 2 | 195 | |||
FQ223794.1 | - | 2 | 212 | |||
FQ223992.1 | - | 2 | 189 | |||
FQ224060.1 | - | 2 | 166 | |||
FQ224189.1 | - | 2 | 210 | |||
FQ224205.1 | - | 2 | 210 | |||
FQ224455.1 | - | 2 | 209 | |||
FQ224486.1 | - | 2 | 166 | |||
FQ224537.1 | - | 2 | 210 | |||
FQ224608.1 | - | 2 | 207 | |||
FQ224652.1 | - | 2 | 171 | |||
FQ224692.1 | - | 2 | 167 |
