siRNA Details
Gene Symbol: Atp5e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139099.1 | NP_620799.1 | 1, 2 | 92 | ||
GenBank mRNA | AF010323.1 | - | 1, 2 | 92 | ||
BC058133.1 | - | 1, 2 | 82 | 6887013 | ||
FQ211630.1 | - | 1, 2 | 114 | |||
FQ211898.1 | - | 1, 2 | 126 | |||
FQ214112.1 | - | 1, 2 | 114 | |||
FQ214935.1 | - | 1, 2 | 117 | |||
FQ215084.1 | - | 1, 2 | 44 | |||
FQ216950.1 | - | 1, 2 | 114 | |||
FQ217027.1 | - | 1, 2 | 78 | |||
FQ217160.1 | - | 1, 2 | 68 | |||
FQ217179.1 | - | 1, 2 | 117 | |||
FQ217559.1 | - | 1, 2 | 67 | |||
FQ217635.1 | - | 1, 2 | 73 | |||
FQ218023.1 | - | 1, 2 | 73 | |||
FQ218046.1 | - | 1, 2 | 73 | |||
FQ222358.1 | - | 1, 2 | 101 | |||
FQ223590.1 | - | 1, 2 | 126 | |||
FQ223763.1 | - | 1, 2 | 114 | |||
FQ223780.1 | - | 1, 2 | 102 | |||
FQ223794.1 | - | 1, 2 | 119 | |||
FQ223992.1 | - | 1, 2 | 96 | |||
FQ224060.1 | - | 1, 2 | 73 | |||
FQ224189.1 | - | 1, 2 | 117 | |||
FQ224205.1 | - | 1, 2 | 117 | |||
FQ224455.1 | - | 1, 2 | 116 | |||
FQ224486.1 | - | 1, 2 | 73 | |||
FQ224537.1 | - | 1, 2 | 117 | |||
FQ224608.1 | - | 1, 2 | 114 | |||
FQ224652.1 | - | 1, 2 | 78 | |||
FQ224692.1 | - | 1, 2 | 74 |
