siRNA Details
| Gene Symbol: Dmgdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139102.2 | NP_620802.2 | 12 | 1958 | ||
| XM_008760629.1 | XP_008758851.1 | 12 | 1973 | |||
| XM_008760630.1 | XP_008758852.1 | 12 | 1973 | |||
| XM_008760631.1 | XP_008758853.1 | 12 | 1973 | |||
| XM_008760632.1 | XP_008758854.1 | 12 | 1973 | |||
| GenBank mRNA | BC085697.1 | - | 12 | 1958 | 7108858 | |
| X55995.1 | - | 12 | 1926 | |||

Dmgdh
Genomic Map