siRNA Details
| Gene Symbol: Dmgdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139102.2 | NP_620802.2 | 16 | 2537 | ||
| GenBank mRNA | BC085697.1 | - | 16 | 2537 | 7108858 | |
| X55995.1 | - | 16 | 2505 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dmgdh| Gene Symbol: Dmgdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139102.2 | NP_620802.2 | 16 | 2537 | ||
| GenBank mRNA | BC085697.1 | - | 16 | 2537 | 7108858 | |
| X55995.1 | - | 16 | 2505 | |||