siRNA Details
| Gene Symbol: Atp5d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139106.1 | NP_620806.1 | 4 | 498 | ||
| XM_006240845.2 | XP_006240907.1 | 5 | 1013 | |||
| GenBank mRNA | BC161836.1 | - | 4 | 2158 | 9098303 | |
| FQ214102.1 | - | 706 | ||||
| U00926.1 | - | 4 | 498 | |||

Atp5d
Genomic Map