siRNA Details
Gene Symbol: Cklf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139111.1 | NP_620811.1 | 3 | 480 | ||
GenBank mRNA | AF253064.2 | - | 2 | 321 | ||
AF253065.2 | - | 3 | 480 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cklf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139111.1 | NP_620811.1 | 3 | 480 | ||
GenBank mRNA | AF253064.2 | - | 2 | 321 | ||
AF253065.2 | - | 3 | 480 |