siRNA Details
| Gene Symbol: Cklf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139111.1 | NP_620811.1 | 4 | 572 | ||
| GenBank mRNA | AF253064.2 | - | 3 | 413 | ||
| AF253065.2 | - | 4 | 572 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cklf| Gene Symbol: Cklf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139111.1 | NP_620811.1 | 4 | 572 | ||
| GenBank mRNA | AF253064.2 | - | 3 | 413 | ||
| AF253065.2 | - | 4 | 572 | |||