siRNA Details
| Gene Symbol: Nr2f6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139113.1 | NP_620813.1 | 2 | 505 | ||
| GenBank mRNA | AF003926.1 | - | 2 | 505 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nr2f6| Gene Symbol: Nr2f6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139113.1 | NP_620813.1 | 2 | 505 | ||
| GenBank mRNA | AF003926.1 | - | 2 | 505 | ||