siRNA Details
| Gene Symbol: Prr21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139184.1 | NP_631923.1 | 2 | 202 | ||
| XM_008763356.1 | XP_008761578.1 | 2 | 201 | |||
| GenBank mRNA | L08134.1 | - | 2 | 202 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prr21| Gene Symbol: Prr21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139184.1 | NP_631923.1 | 2 | 202 | ||
| XM_008763356.1 | XP_008761578.1 | 2 | 201 | |||
| GenBank mRNA | L08134.1 | - | 2 | 202 | ||